ENA066595989 Enamine

4,6,7-trimethyl-1H-indole-2-carboxylic acid

  • MDL number MFCD02664504
  • Cas Number : 383133-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227...229
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15118

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15118
Availability details for
Quantity
  • Availability
  • Add to Cart