ENA066595995 Enamine

2-(carbamoylmethoxy)benzoic acid

  • MDL number MFCD04508187
  • Cas Number : 56424-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227-229
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15368
Availability details for
Quantity
  • Availability
  • Add to Cart