ENA066596023 Enamine

2-{[(3,4-dimethylphenyl)sulfonyl]amino}-3-hydroxybutanoic acid

  • MDL number MFCD07323195
  • Cas Number : 1008068-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15782

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15782
Availability details for
Quantity
  • Availability
  • Add to Cart