ENA066596043 Enamine

2-({[(propan-2-yl)carbamoyl]methyl}sulfanyl)benzoic acid

  • MDL number MFCD06364964
  • Cas Number : 793727-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-16509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16509
Availability details for
Quantity
  • Availability
  • Add to Cart