ENA066596045 Enamine

4-amino-5-[(4-chlorophenoxy)methyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00128286
  • Cas Number : 4413-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-16514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16514
Availability details for
Quantity
  • Availability
  • Add to Cart