ENA066596056 Enamine

5-(4-fluorophenyl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD00704683
  • Cas Number : 428-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16728

Safety Information

msds https://www.enaminestore.com/catalog/EN300-16728
Availability details for
Quantity
  • Availability
  • Add to Cart