ENA066596060 Enamine

{[4-(difluoromethoxy)benzoyl]amino}acetic acid

  • MDL number MFCD01918125
  • Cas Number : 304685-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16803

Safety Information

msds https://www.enaminestore.com/catalog/EN300-16803
Availability details for
Quantity
  • Availability
  • Add to Cart