ENA066596062 Enamine

3-[4-(piperidine-1-sulfonyl)phenyl]propanoic acid

  • MDL number MFCD02708102
  • Cas Number : 327091-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-16846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16846
Availability details for
Quantity
  • Availability
  • Add to Cart