ENA066596080 Enamine

2,2-dioxo-3H,4H-2lambda6-pyrido[2,1-c][1,2,4]thiadiazine-9-carboxylic acid

  • MDL number MFCD03964729
  • Cas Number : 565165-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-150
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-17121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-17121
Availability details for
Quantity
  • Availability
  • Add to Cart