ENA066596113 Enamine

2-(chloromethyl)-1-methoxy-4-nitrobenzene

  • MDL number MFCD06359953
  • Cas Number : 93-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-17781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-17781
Availability details for
Quantity
  • Availability
  • Add to Cart