ENA066596116 Enamine

4-methyl-5-[2-(4-methylphenyl)quinolin-4-yl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03943432
  • Cas Number : 588673-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-17807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-17807
Availability details for
Quantity
  • Availability
  • Add to Cart