ENA066596122 Enamine

5-{[3-(morpholin-4-yl)propyl]amino}-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD06366763
  • Cas Number : 794554-82-8
  • Molecular Weight : 260.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-17846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-17846
Availability details for
Quantity
  • Availability
  • Add to Cart