ENA066596123 Enamine

2-bromo-1-[2-(difluoromethoxy)-5-methylphenyl]ethan-1-one

  • MDL number MFCD06368763
  • Cas Number : 795290-97-0
  • Molecular Weight : 279.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-17859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-17859
Availability details for
Quantity
  • Availability
  • Add to Cart