ENA066596126 Enamine

5-[4-(diethylamino)phenyl]-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD06370782
  • Cas Number : 796067-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-149
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-17869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-17869
Availability details for
Quantity
  • Availability
  • Add to Cart