ENA066596139 Enamine

3-bromo-5-ethoxy-4-propoxybenzaldehyde

  • MDL number MFCD01169261
  • Cas Number : 834908-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-18314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-18314
Availability details for
Quantity
  • Availability
  • Add to Cart