ENA066596143 Enamine

7-chloro-3-(propan-2-yl)-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD08271811
  • Cas Number : 422526-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18460

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-18460
Availability details for
Quantity
  • Availability
  • Add to Cart