ENA066596151 Enamine

2-chloro-N'-[(4-chlorophenoxy)acetyl]acetohydrazide

  • MDL number MFCD01121440
  • Cas Number : 113603-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18490

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18490
Availability details for
Quantity
  • Availability
  • Add to Cart