ENA066596155 Enamine

2-[(2-methoxyphenyl)amino]pyridine-3-carboxylic acid

  • MDL number MFCD05241565
  • Cas Number : 114501-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-186
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-18543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-18543
Availability details for
Quantity
  • Availability
  • Add to Cart