ENA066596162 Enamine

4-(methylsulfanyl)-2-[(5-nitrofuran-2-yl)formamido]butanoic acid

  • MDL number MFCD06674594
  • Cas Number : 1008068-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-18614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-18614
Availability details for
Quantity
  • Availability
  • Add to Cart