ENA066596177 Enamine

5-(ethylsulfonyl)-2-methyl-1,3-benzoxazole

  • MDL number MFCD07325229
  • Cas Number : 847837-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18904

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18904
Availability details for
Quantity
  • Availability
  • Add to Cart