ENA066596207 Enamine

4-methyl-6-(piperazin-1-yl)-2-(propan-2-yl)pyrimidine

  • MDL number MFCD07400493
  • Cas Number : 873208-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-41
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21799

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21799
Availability details for
Quantity
  • Availability
  • Add to Cart