ENA066596212 Enamine

6-amino-1,3-dioxaindane-5-carboxylic acid

  • MDL number MFCD00016650
  • Cas Number : 20332-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-15140

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15140
Availability details for
Quantity
  • Availability
  • Add to Cart