ENA066596222 Enamine

5-(4-ethoxyphenyl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD00440311
  • Cas Number : 68524-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-191
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-21814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21814
Availability details for
Quantity
  • Availability
  • Add to Cart