ENA066596229 Enamine

5-[(4-fluorophenyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD07366447
  • Cas Number : 14731-24-9
  • Molecular Weight : 227.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-186
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-15011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15011
Availability details for
Quantity
  • Availability
  • Add to Cart