ENA066596232 Enamine

5-bromo-1,3-diazinane-2,4-dione

  • MDL number MFCD00023158
  • Cas Number : 1193-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-193
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21829
Availability details for
Quantity
  • Availability
  • Add to Cart