ENA066596234 Enamine

5-(4-bromophenyl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD00455252
  • Cas Number : 6320-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 262...264
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21832

Safety Information

msds https://www.enaminestore.com/catalog/EN300-21832
Availability details for
Quantity
  • Availability
  • Add to Cart