ENA066596241 Enamine

2-chloro-N-[2-(piperidin-1-yl)-5-(trifluoromethyl)phenyl]acetamide

  • MDL number MFCD07689450
  • Cas Number : 496012-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21849
Availability details for
Quantity
  • Availability
  • Add to Cart