ENA066596248 Enamine

2-(5-thien-3-yl-2H-tetrazol-2-yl)butanoic acid

  • MDL number MFCD07681943
  • Cas Number : 871909-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 263...265
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15159

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15159
Availability details for
Quantity
  • Availability
  • Add to Cart