ENA068330846 Enamine

N-(4-bromobenzyl)-2-chloro-N-methylacetamide

  • MDL number MFCD08262487
  • Cas Number : 879362-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-22355
Availability details for
Quantity
  • Availability
  • Add to Cart