ENA068330854 Enamine

5,6-dibromo-1H-indole-3-carboxylic acid

  • MDL number MFCD08271830
  • Cas Number : 857809-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23187

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23187
Availability details for
Quantity
  • Availability
  • Add to Cart