ENA068330863 Enamine

2-(4-fluorophenyl)-2-(piperazin-1-yl)acetonitrile

  • MDL number MFCD07841699
  • Cas Number : 875858-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-64
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23431

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23431
Availability details for
Quantity
  • Availability
  • Add to Cart