ENA068330864 Enamine

2-(2-chloroacetamido)-N-(2-phenylethyl)benzamide

  • MDL number MFCD07838341
  • Cas Number : 117507-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23440
Availability details for
Quantity
  • Availability
  • Add to Cart