ENA068330875 Enamine

2-(chloromethyl)-6-(4-methoxyphenyl)thieno[3,2-d]pyrimidin-4(1H)-one

  • MDL number MFCD07838298
  • Cas Number : 875156-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23503

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23503
Availability details for
Quantity
  • Availability
  • Add to Cart