ENA068330881 Enamine

4,5-dichloro-2-[2-(4-chlorophenoxy)ethyl]-2,3-dihydropyridazin-3-one

  • MDL number MFCD07838432
  • Cas Number : 875164-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23528
Availability details for
Quantity
  • Availability
  • Add to Cart