ENA068330897 Enamine

{2-[(4-chlorophenoxy)methyl]-1,3-thiazol-4-yl}acetic acid

  • MDL number MFCD06858085
  • Cas Number : 876866-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23569

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23569
Availability details for
Quantity
  • Availability
  • Add to Cart