ENA068330898 Enamine

2-[(4-chlorophenoxy)methyl]-4-methyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD06858122
  • Cas Number : 187998-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23570
Availability details for
Quantity
  • Availability
  • Add to Cart