ENA068330905 Enamine

3-(chloromethyl)-5-(4-fluorophenyl)-1,2,4-oxadiazole

  • MDL number MFCD07838531
  • Cas Number : 166179-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-53
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23590

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23590
Availability details for
Quantity
  • Availability
  • Add to Cart