ENA068330906 Enamine

2-(2-phenyl-1,3-oxazol-4-yl)acetic acid

  • MDL number MFCD07838437
  • Cas Number : 22086-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23594
Availability details for
Quantity
  • Availability
  • Add to Cart