ENA068330908 Enamine

1-(morpholin-4-yl)-2-(piperazin-1-yl)ethane-1,2-dione

  • MDL number MFCD07838439
  • Cas Number : 875164-29-7
  • Molecular Weight : 227.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-23599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23599
Availability details for
Quantity
  • Availability
  • Add to Cart