ENA068330914 Enamine

2-{2,4-dioxo-1,3-diazaspiro[4.4]nonan-3-yl}ethane-1-sulfonyl fluoride

  • MDL number MFCD08271853
  • Cas Number : 885527-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23623
Availability details for
Quantity
  • Availability
  • Add to Cart