ENA068330943 Enamine

2-(9-chloro-3,4-dihydro-2H-1,5-benzodioxepin-7-yl)acetic acid

  • MDL number MFCD07691375
  • Cas Number : 874623-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15410
Availability details for
Quantity
  • Availability
  • Add to Cart