ENA068330945 Enamine

1-phenyl-2-[(5-sulfanyl-1,3,4-thiadiazol-2-yl)sulfanyl]ethan-1-one

  • MDL number MFCD03102872
  • Cas Number : 866144-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187-188
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-15385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15385
Availability details for
Quantity
  • Availability
  • Add to Cart