ENA068330948 Enamine

3-[3-(trifluoromethyl)phenoxy]benzoic acid

  • MDL number MFCD06203216
  • Cas Number : 127389-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15413

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15413
Availability details for
Quantity
  • Availability
  • Add to Cart