ENA068330950 Enamine

1-ethyl-1H-indole-3-carboxylic acid

  • MDL number MFCD04035647
  • Cas Number : 132797-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-134
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23394

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23394
Availability details for
Quantity
  • Availability
  • Add to Cart