ENA068330953 Enamine

2-chloro-N-(1-methylheptyl)acetamide

  • MDL number MFCD07692308
  • Cas Number : 23602-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23408

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23408
Availability details for
Quantity
  • Availability
  • Add to Cart