ENA068330962 Enamine

4-(cyclopropylamino)-2,3-dihydroquinazoline-2-thione

  • MDL number MFCD26143664
  • Molecular Weight : 217.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-23476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23476
Availability details for
Quantity
  • Availability
  • Add to Cart