ENA068330970 Enamine

N-[5-(chloroacetyl)-2-(methylthio)phenyl]acetamide

  • MDL number MFCD07691362
  • Cas Number : 874623-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23484

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23484
Availability details for
Quantity
  • Availability
  • Add to Cart