ENA068330973 Enamine

2,7-dichloro-4H-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD07692319
  • Cas Number : 278614-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222-223
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15420
Availability details for
Quantity
  • Availability
  • Add to Cart