ENA068330995 Enamine

1-(1,3-thiazol-2-yl)piperidine-4-carboxylic acid

  • MDL number MFCD07692337
  • Cas Number : 874623-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15433
Availability details for
Quantity
  • Availability
  • Add to Cart