ENA068331003 Enamine

(2Z)-(hydroxyimino)(4-methyl-1,3-thiazol-2-yl)acetonitrile

  • MDL number MFCD03011718
  • Cas Number : 1164509-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15437

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15437
Availability details for
Quantity
  • Availability
  • Add to Cart